The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020759.2 | NP_065810.2 | 6 - 7 | 492 | 68 |
XM_011521831.2 | XP_011520133.1 | 6 - 7 | 505 | 68 | |
XM_011521832.2 | XP_011520134.1 | 6 - 7 | 502 | 68 | |
XM_011521833.2 | XP_011520135.1 | 5 - 6 | 472 | 68 | |
XM_011521834.2 | XP_011520136.1 | 6 - 7 | 505 | 68 | |
XM_011521835.2 | XP_011520137.1 | 6 - 7 | 505 | 68 | |
XM_011521836.2 | XP_011520138.1 | 6 - 7 | 807 | 68 | |
XM_011521837.2 | XP_011520139.1 | 4 - 5 | 274 | 68 | |
XM_017022439.1 | XP_016877928.1 | 6 - 7 | 505 | 68 | |
XM_017022440.1 | XP_016877929.1 | 5 - 6 | 404 | 68 | |
XM_017022441.1 | XP_016877930.1 | 4 - 5 | 723 | 68 | |
GenBank mRNA | AK122666.1 | - | 4 - 5 | 506 | 68 |
CR627426.1 | - | 6 - 7 | 492 | 68 |