The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301239.1 | NP_001288168.1 | 1 - 2 | 773 | 105 |
NM_006798.4 | NP_006789.3 | 2 - 3 | 835 | 105 | |
GenBank mRNA | AJ006054.1 | - | 1 - 2 | 782 | 105 |
BC142640.1 | - | 1 - 2 | 836 | 105 |