The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318046.1 | NP_001304975.1 | 27 - 28 | 4208 | 152 |
NM_007356.2 | NP_031382.2 | 27 - 28 | 4211 | 152 | |
XM_011515975.1 | XP_011514277.1 | 27 - 28 | 4211 | 152 | |
XM_011515978.1 | XP_011514280.1 | 27 - 28 | 4211 | 152 | |
XM_011515979.2 | XP_011514281.1 | 27 - 28 | 4211 | 152 | |
XM_011515980.2 | XP_011514282.1 | 27 - 28 | 4211 | 152 | |
XM_017011879.1 | XP_016867368.1 | 27 - 28 | 4211 | 152 | |
XM_017011880.1 | XP_016867369.1 | 27 - 28 | 4211 | 152 | |
GenBank mRNA | AF028816.1 | - | 27 - 28 | 4207 | 152 |
AK301777.1 | - | 6 - 7 | 1203 | 152 | |
BC045172.2 | - | 6 - 7 | 1205 | 152 | |
BC140804.1 | - | 27 - 28 | 4198 | 152 | |
BC144591.1 | - | 27 - 28 | 4198 | 152 |