The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271969.1 | NP_001258898.1 | 9 - 10 | 1716 | 79 |
NM_001271970.1 | NP_001258899.1 | 9 - 10 | 1644 | 79 | |
NM_001271971.1 | NP_001258900.1 | 9 - 10 | 1559 | 79 | |
NM_001271972.1 | NP_001258901.1 | 9 - 10 | 1673 | 79 | |
NM_007355.3 | NP_031381.2 | 9 - 10 | 1703 | 79 | |
GenBank mRNA | AF275719.1 | - | 7 - 8 | 1189 | 79 |
AK025370.1 | - | 6 - 7 | 1006 | 79 | |
AK130494.1 | - | 1 - 2 | 153 | 79 | |
AK290734.1 | - | 4 - 5 | 822 | 79 | |
AK294643.1 | - | 9 - 10 | 1530 | 79 | |
AK297367.1 | - | 9 - 10 | 1446 | 79 | |
AK312255.1 | - | 9 - 10 | 1564 | 79 | |
AL136543.1 | - | 9 - 10 | 1539 | 79 | |
AY359878.1 | - | 9 - 10 | 1577 | 79 | |
BC004928.1 | - | 9 - 10 | 1549 | 79 | |
BC009206.2 | - | 7 - 8 | 1243 | 79 | |
BC012807.2 | - | 9 - 10 | 1549 | 79 | |
BC014485.1 | - | 9 - 10 | 1544 | 79 | |
BC016753.2 | - | 9 - 10 | 1562 | 79 | |
BC068474.1 | - | 9 - 10 | 1567 | 79 | |
BX649079.1 | - | 9 - 10 | 1566 | 79 | |
M16660.1 | - | 9 - 10 | 1549 | 79 |