Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
Chromosome Location
Chr.7: 43112580 - 43566001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287059.1 NP_001273988.1 17 - 18 3710 68
NM_015052.4 NP_055867.3 19 - 20 4047 68
XM_005249665.3 XP_005249722.1 18 - 19 3829 68
XM_006715670.3 XP_006715733.1 17 - 18 3727 68
XM_006715671.3 XP_006715734.1 17 - 18 3724 68
XM_006715673.3 XP_006715736.1 16 - 17 3622 68
XM_011515220.2 XP_011513522.1 18 - 19 3620 68
XM_011515222.2 XP_011513524.1 17 - 18 3761 68
XM_011515223.2 XP_011513525.1 17 - 18 3562 68
XM_011515224.2 XP_011513526.1 17 - 18 4042 68
XM_011515225.2 XP_011513527.2 15 - 16 3079 68
XM_011515226.2 XP_011513528.1 14 - 15 3303 68
XM_017011882.1 XP_016867371.1 17 - 18 4069 68
XM_017011883.1 XP_016867372.1 18 - 19 4006 68
XM_017011884.1 XP_016867373.1 17 - 18 4066 68
XM_017011885.1 XP_016867374.1 19 - 20 4002 68
XM_017011886.1 XP_016867375.1 18 - 19 3831 68
XM_017011887.1 XP_016867376.1 16 - 17 3967 68
XM_017011888.1 XP_016867377.1 16 - 17 3964 68
XM_017011889.1 XP_016867378.1 17 - 18 4021 68
XM_017011890.1 XP_016867379.1 18 - 19 3831 68
GenBank mRNA AB002320.2 - 19 - 20 4047 68
AB048365.1 - 16 - 17 3399 68
AK294918.1 - 17 - 18 3664 68
BC151227.1 - 19 - 20 4047 68

View Full Product Details