Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
Chromosome Location
Chr.7: 43112580 - 43566001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287059.1 NP_001273988.1 20 - 21 4021 89
NM_015052.4 NP_055867.3 22 - 23 4358 89
XM_005249665.3 XP_005249722.1 21 - 22 4140 89
XM_006715670.3 XP_006715733.1 20 - 21 4038 89
XM_006715671.3 XP_006715734.1 20 - 21 4035 89
XM_006715673.3 XP_006715736.1 19 - 20 3933 89
XM_011515220.2 XP_011513522.1 21 - 22 3931 89
XM_011515222.2 XP_011513524.1 20 - 21 4072 89
XM_011515223.2 XP_011513525.1 20 - 21 3873 89
XM_011515224.2 XP_011513526.1 20 - 21 4353 89
XM_011515225.2 XP_011513527.2 18 - 19 3390 89
XM_011515226.2 XP_011513528.1 17 - 18 3614 89
XM_017011882.1 XP_016867371.1 20 - 21 4380 89
XM_017011883.1 XP_016867372.1 21 - 22 4317 89
XM_017011884.1 XP_016867373.1 20 - 21 4377 89
XM_017011885.1 XP_016867374.1 22 - 23 4313 89
XM_017011886.1 XP_016867375.1 21 - 22 4142 89
XM_017011887.1 XP_016867376.1 19 - 20 4278 89
XM_017011888.1 XP_016867377.1 19 - 20 4275 89
XM_017011889.1 XP_016867378.1 20 - 21 4332 89
XM_017011890.1 XP_016867379.1 21 - 22 4142 89
GenBank mRNA AB002320.2 - 22 - 23 4358 89
AB048365.1 - 19 - 20 3710 89
AK294918.1 - 20 - 21 3975 89
BC151227.1 - 22 - 23 4358 89

View Full Product Details