The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012720.1 | NP_001012738.1 | 5 - 6 | 667 | 73 |
NM_002921.3 | NP_002912.2 | 5 - 6 | 679 | 73 | |
GenBank mRNA | BC011349.1 | - | 5 - 6 | 653 | 73 |
BX641033.1 | - | 4 - 5 | 1841 | 73 | |
KU178307.1 | - | 5 - 6 | 641 | 73 | |
U14910.1 | - | 5 - 6 | 667 | 73 |