The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172509.1 | NP_001165980.1 | 10 - 11 | 2557 | 82 |
NM_001172517.1 | NP_001165988.1 | 11 - 12 | 2153 | 82 | |
NM_015265.3 | NP_056080.1 | 11 - 12 | 2133 | 82 | |
XM_005246396.2 | XP_005246453.1 | 9 - 10 | 1788 | 82 | |
XM_006712372.2 | XP_006712435.1 | 10 - 11 | 1903 | 82 | |
XM_011510840.2 | XP_011509142.1 | 11 - 12 | 2127 | 82 | |
XM_017003656.1 | XP_016859145.1 | 10 - 11 | 1788 | 82 | |
GenBank mRNA | AB028957.1 | - | 10 - 11 | 1826 | 82 |
AB209376.1 | - | 11 - 12 | 2153 | 82 | |
AK056638.1 | - | 10 - 11 | 2558 | 82 | |
AK291463.1 | - | 11 - 12 | 2139 | 82 | |
BC098136.1 | - | 9 - 10 | 1791 | 82 | |
BC099723.1 | - | 9 - 10 | 1791 | 82 | |
BC103492.1 | - | 6 - 7 | 1437 | 82 | |
BC103500.1 | - | 9 - 10 | 1791 | 82 |