The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174081.1 | NP_001167552.1 | 3 - 4 | 440 | 85 |
NM_001174082.1 | NP_001167553.1 | 3 - 4 | 471 | 85 | |
NM_001174083.1 | NP_001167554.1 | 4 - 5 | 402 | 85 | |
NM_017852.3 | NP_060322.1 | 3 - 4 | 471 | 85 | |
GenBank mRNA | AF298547.1 | - | 3 - 4 | 388 | 85 |
AF310106.1 | - | 3 - 4 | 416 | 85 | |
AF464764.1 | - | 3 - 4 | 371 | 85 | |
AK000517.1 | - | 3 - 4 | 416 | 85 | |
AK025952.1 | - | 3 - 4 | 380 | 85 | |
AK223253.1 | - | 3 - 4 | 380 | 85 | |
AK223269.1 | - | 3 - 4 | 380 | 85 | |
AK292681.1 | - | 3 - 4 | 408 | 85 | |
AK302989.1 | - | 4 - 5 | 371 | 85 | |
BC001039.2 | - | 2 - 3 | 351 | 85 | |
BC003592.1 | - | 3 - 4 | 395 | 85 | |
BC039269.1 | - | 3 - 4 | 440 | 85 | |
DA856893.1 | - | 3 - 4 | 440 | 85 | |
DB046128.1 | - | 3 - 4 | 471 | 85 |