The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300868.1 | NP_001287797.1 | 8 - 9 | 1081 | 76 |
NM_001300869.1 | NP_001287798.1 | 8 - 9 | 1046 | 76 | |
NM_001532.2 | NP_001523.2 | 6 - 7 | 880 | 76 | |
NR_125343.1 | 6 - 7 | 880 | 76 | ||
XM_011544974.1 | XP_011543276.1 | 8 - 9 | 1031 | 76 | |
XM_017017630.1 | XP_016873119.1 | 6 - 7 | 1234 | 76 | |
XM_017017631.1 | XP_016873120.1 | 6 - 7 | 1234 | 76 | |
XM_017017632.1 | XP_016873121.1 | 8 - 9 | 1031 | 76 | |
XM_017017633.1 | XP_016873122.1 | 6 - 7 | 1234 | 76 | |
XM_017017634.1 | XP_016873123.1 | 6 - 7 | 1234 | 76 | |
XM_017017635.1 | XP_016873124.1 | 6 - 7 | 1234 | 76 | |
XM_017017636.1 | XP_016873125.1 | 6 - 7 | 1234 | 76 | |
XM_017017637.1 | XP_016873126.1 | 6 - 7 | 1234 | 76 | |
XM_017017638.1 | XP_016873127.1 | 4 - 5 | 404 | 76 | |
GenBank mRNA | AF029358.1 | - | 6 - 7 | 654 | 76 |
AF034102.1 | - | 6 - 7 | 888 | 76 | |
AF401235.1 | - | 6 - 7 | 888 | 76 | |
AK057041.1 | - | 7 - 8 | 871 | 76 | |
BC011387.1 | - | 7 - 8 | 761 | 76 | |
BC093634.1 | - | 6 - 7 | 700 | 76 | |
BC143335.1 | - | 6 - 7 | 700 | 76 | |
X86681.1 | - | 6 - 7 | 646 | 76 |