Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAR1
Entrez Gene ID
9564
Gene Name
BCAR1, Cas family scaffolding protein
Gene Aliases
CAS, CAS1, CASS1, CRKAS, P130Cas
Chromosome Location
Chr.16: 75229030 - 75268053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170714.1 NP_001164185.1 5 - 6 1334 52
NM_001170715.1 NP_001164186.1 4 - 5 1039 52
NM_001170716.1 NP_001164187.1 4 - 5 996 52
NM_001170718.1 NP_001164189.1 4 - 5 1083 52
NM_001170719.1 NP_001164190.1 4 - 5 1173 52
NM_001170720.1 NP_001164191.1 5 - 6 744 52
NM_001170721.1 NP_001164192.1 3 - 4 1271 52
NM_014567.3 NP_055382.2 4 - 5 1058 52
XM_005256258.4 XP_005256315.1 4 - 5 1258 52
XM_005256259.4 XP_005256316.1 4 - 5 4170 52
XM_005256260.4 XP_005256317.1 4 - 5 1179 52
XM_011523475.2 XP_011521777.1 4 - 5 1207 52
XM_017023897.1 XP_016879386.1 4 - 5 1765 52
XM_017023898.1 XP_016879387.1 4 - 5 1765 52
GenBank mRNA AB040024.1 - 4 - 5 1053 52
AF218451.1 - 4 - 5 1036 52
AJ242987.2 - 4 - 5 1033 52
AK027608.1 - 4 - 5 1083 52
AK074958.1 - 3 - 4 468 52
AK124526.1 - 4 - 5 2478 52
AK124815.1 - 4 - 5 1042 52
AK124844.1 - 3 - 4 849 52
AK293808.1 - 5 - 6 1334 52
AK294513.1 - 4 - 5 1089 52
AK295809.1 - 4 - 5 968 52
AK302617.1 - 5 - 6 744 52
AK314722.1 - 4 - 5 1058 52
BC062556.1 - 3 - 4 913 52

View Full Product Details