Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
319089
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
C14orf25, NCRNA00291
Chromosome Location
Chr.14: 37595199 - 37842628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX161415.1 - 5 - 6 636 92

View Full Product Details