The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007361.3 | NP_031387.3 | 3 - 4 | 1008 | 87 |
XM_005267405.4 | XP_005267462.1 | 3 - 4 | 1011 | 87 | |
XM_005267406.4 | XP_005267463.1 | 3 - 4 | 1011 | 87 | |
XM_005267407.4 | XP_005267464.1 | 3 - 4 | 1011 | 87 | |
GenBank mRNA | AJ223500.1 | - | 3 - 4 | 774 | 87 |
AK291652.1 | - | 3 - 4 | 836 | 87 | |
AK300462.1 | - | 4 - 5 | 646 | 87 | |
BX648241.1 | - | 4 - 5 | 781 | 87 | |
D86425.1 | - | 3 - 4 | 777 | 87 |