Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX10
Entrez Gene ID
347853
Gene Name
T-box 10
Gene Aliases
TBX13, TBX7
Chromosome Location
Chr.11: 67631303 - 67639560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005995.4 NP_005986.2 1 - 2 92 53
GenBank mRNA AY229977.1 - 1 - 2 93 53
BC113485.1 - 1 - 2 86 53
BC113487.1 - 1 - 2 86 53

View Full Product Details