The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005386.2 | NP_001005386.1 | 1 - 2 | 262 | 70 |
NM_005722.3 | NP_005713.1 | 1 - 2 | 262 | 70 | |
GenBank mRNA | AF006082.1 | - | 1 - 2 | 121 | 70 |
AK295157.1 | - | 1 - 2 | 212 | 70 | |
AK301634.1 | - | 1 - 2 | 124 | 70 | |
AK315205.1 | - | 1 - 2 | 111 | 70 | |
BC014546.1 | - | 1 - 2 | 128 | 70 | |
BX491447.1 | - | 1 - 2 | 167 | 70 | |
DC404298.1 | - | 1 - 2 | 262 | 70 |