The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322270.1 | NP_001309199.1 | 2 - 3 | 233 | 143 |
NM_001322271.1 | NP_001309200.1 | 2 - 3 | 233 | 143 | |
NM_001322272.1 | NP_001309201.1 | 2 - 3 | 438 | 143 | |
NM_017437.2 | NP_059133.1 | 2 - 3 | 233 | 143 | |
GenBank mRNA | AK001627.2 | - | 2 - 3 | 210 | 143 |
AK001912.1 | - | 2 - 3 | 203 | 143 | |
AU132572.1 | - | 2 - 3 | 210 | 143 | |
BC070095.1 | - | 2 - 3 | 203 | 143 | |
DB081880.1 | - | 2 - 3 | 233 | 143 |