The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006606.2 | NP_006597.2 | 1 - 2 | 182 | 82 |
GenBank mRNA | AF039564.1 | - | 1 - 2 | 106 | 82 |
AF237576.1 | - | 1 - 2 | 136 | 82 | |
AL832411.1 | - | 1 - 2 | 130 | 82 | |
BC015938.1 | - | 1 - 2 | 182 | 82 |