The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190463.1 | NP_001177392.1 | 8 - 9 | 1091 | 121 |
NM_001902.5 | NP_001893.2 | 9 - 10 | 1187 | 121 | |
NM_153742.4 | NP_714964.2 | 8 - 9 | 1055 | 121 | |
XM_005270509.3 | XP_005270566.1 | 9 - 10 | 1207 | 121 | |
XM_017000416.1 | XP_016855905.1 | 6 - 7 | 911 | 121 | |
GenBank mRNA | AK223376.1 | - | 9 - 10 | 1160 | 121 |
AK303946.1 | - | 8 - 9 | 1068 | 121 | |
BC015807.2 | - | 9 - 10 | 1133 | 121 | |
BT006882.1 | - | 9 - 10 | 989 | 121 | |
S52028.1 | - | 8 - 9 | 890 | 121 | |
S52784.1 | - | 9 - 10 | 1022 | 121 |