The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178111.1 | NP_001171582.1 | 3 - 4 | 280 | 79 |
NM_001178112.1 | NP_001171583.1 | 3 - 4 | 247 | 79 | |
NM_138967.3 | NP_620417.1 | 2 - 3 | 197 | 79 | |
NR_033660.1 | 2 - 3 | 197 | 79 | ||
XM_006720420.1 | XP_006720483.1 | 2 - 3 | 356 | 79 | |
XM_006720421.1 | XP_006720484.1 | 3 - 4 | 238 | 79 | |
XM_006720422.3 | XP_006720485.1 | 3 - 4 | 206 | 79 | |
XM_017021989.1 | XP_016877478.1 | 2 - 3 | 346 | 79 | |
GenBank mRNA | AB105192.1 | - | 2 - 3 | 168 | 79 |
AF495715.1 | - | 2 - 3 | 149 | 79 | |
AK056438.1 | - | 3 - 4 | 222 | 79 | |
AK126940.1 | - | 2 - 3 | 197 | 79 | |
AK127480.1 | - | 3 - 4 | 217 | 79 | |
AK301505.1 | - | 2 - 3 | 159 | 79 | |
AL834226.1 | - | 2 - 3 | 168 | 79 | |
BC024700.1 | - | 1 - 2 | 40 | 79 | |
CR936761.1 | - | 2 - 3 | 130 | 79 | |
DA810757.1 | - | 2 - 3 | 197 | 79 |