The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002031.2 | NP_002022.1 | 2 - 3 | 905 | 114 |
XM_005266880.4 | XP_005266937.1 | 3 - 4 | 977 | 114 | |
XM_005266881.2 | XP_005266938.1 | 3 - 4 | 894 | 114 | |
XM_005266882.4 | XP_005266939.1 | 4 - 5 | 1206 | 114 | |
XM_011535653.2 | XP_011533955.1 | 4 - 5 | 1209 | 114 | |
XM_011535654.2 | XP_011533956.1 | 3 - 4 | 991 | 114 | |
XM_011535655.2 | XP_011533957.1 | 3 - 4 | 988 | 114 | |
XM_011535656.2 | XP_011533958.1 | 2 - 3 | 362 | 114 | |
XM_017010645.1 | XP_016866134.1 | 3 - 4 | 1095 | 114 | |
GenBank mRNA | AK313314.1 | - | 2 - 3 | 985 | 114 |
BC012916.1 | - | 2 - 3 | 750 | 114 | |
U00803.1 | - | 2 - 3 | 905 | 114 | |
U22322.1 | - | 2 - 3 | 867 | 114 |