The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002031.2 | NP_002022.1 | 3 - 4 | 1080 | 83 |
XM_005266880.4 | XP_005266937.1 | 4 - 5 | 1152 | 83 | |
XM_005266881.2 | XP_005266938.1 | 4 - 5 | 1069 | 83 | |
XM_005266882.4 | XP_005266939.1 | 5 - 6 | 1381 | 83 | |
XM_011535653.2 | XP_011533955.1 | 5 - 6 | 1384 | 83 | |
XM_011535654.2 | XP_011533956.1 | 4 - 5 | 1166 | 83 | |
XM_011535655.2 | XP_011533957.1 | 4 - 5 | 1163 | 83 | |
XM_011535656.2 | XP_011533958.1 | 3 - 4 | 537 | 83 | |
XM_017010645.1 | XP_016866134.1 | 4 - 5 | 1270 | 83 | |
GenBank mRNA | AK302264.1 | - | 2 - 3 | 255 | 83 |
AK313314.1 | - | 3 - 4 | 1160 | 83 | |
BC012916.1 | - | 3 - 4 | 925 | 83 | |
U00803.1 | - | 3 - 4 | 1080 | 83 | |
U22322.1 | - | 3 - 4 | 1042 | 83 |