Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRK
Entrez Gene ID
2444
Gene Name
fyn related Src family tyrosine kinase
Gene Aliases
GTK, PTK5, RAK
Chromosome Location
Chr.6: 115941031 - 116100739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002031.2 NP_002022.1 4 - 5 1246 74
XM_005266880.4 XP_005266937.1 5 - 6 1318 74
XM_005266881.2 XP_005266938.1 5 - 6 1235 74
XM_005266882.4 XP_005266939.1 6 - 7 1547 74
XM_011535653.2 XP_011533955.1 6 - 7 1550 74
XM_011535654.2 XP_011533956.1 5 - 6 1332 74
XM_011535655.2 XP_011533957.1 5 - 6 1329 74
XM_011535656.2 XP_011533958.1 4 - 5 703 74
XM_017010645.1 XP_016866134.1 5 - 6 1436 74
GenBank mRNA AK302264.1 - 3 - 4 421 74
AK313314.1 - 4 - 5 1326 74
BC012916.1 - 4 - 5 1091 74
U00803.1 - 4 - 5 1246 74
U22322.1 - 4 - 5 1208 74

View Full Product Details