The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282862.1 | NP_001269791.1 | 6 - 7 | 866 | 63 |
NM_145313.3 | NP_660356.2 | 6 - 7 | 838 | 63 | |
XM_005271809.3 | XP_005271866.1 | 5 - 6 | 716 | 63 | |
XM_011539500.2 | XP_011537802.1 | 5 - 6 | 826 | 63 | |
GenBank mRNA | AK095136.1 | - | 6 - 7 | 866 | 63 |
AK127432.1 | - | 6 - 7 | 3264 | 63 |