The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000182.4 | NP_000173.2 | 2 - 3 | 243 | 70 |
GenBank mRNA | AK299258.1 | - | 2 - 3 | 129 | 70 |
AK302532.1 | - | 2 - 3 | 124 | 70 | |
AK313027.1 | - | 2 - 3 | 136 | 70 | |
BC009235.2 | - | 2 - 3 | 138 | 70 | |
CN264228.1 | - | 2 - 3 | 243 | 70 | |
D16480.1 | - | 2 - 3 | 140 | 70 | |
FJ626228.1 | - | 2 - 3 | 113 | 70 | |
GU727642.1 | - | 2 - 3 | 138 | 70 | |
U04627.1 | - | 2 - 3 | 139 | 70 |