Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HADHA
Entrez Gene ID
3030
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
Chromosome Location
Chr.2: 26190635 - 26244726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000182.4 NP_000173.2 4 - 5 446 74
GenBank mRNA AK299258.1 - 4 - 5 332 74
AK313027.1 - 4 - 5 339 74
BC009235.2 - 4 - 5 341 74
D16480.1 - 4 - 5 343 74
GU727642.1 - 4 - 5 341 74
U04627.1 - 4 - 5 342 74

View Full Product Details