Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLL2
Entrez Gene ID
7093
Gene Name
tolloid like 2
Gene Aliases
-
Chromosome Location
Chr.10: 96364606 - 96513926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012465.3 NP_036597.1 5 - 6 880 73
GenBank mRNA AB023149.2 - 5 - 6 830 73
AF059516.1 - 5 - 6 1003 73
AK026106.1 - 4 - 5 716 73
BC013871.2 - 5 - 6 840 73
BC112341.1 - 5 - 6 763 73
BC112366.1 - 5 - 6 763 73
BC113577.1 - 5 - 6 696 73

View Full Product Details