The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143682.1 | NP_001137154.1 | 12 - 13 | 1677 | 60 |
NM_020898.2 | NP_065949.1 | 13 - 14 | 1932 | 60 | |
NR_026554.1 | 12 - 13 | 1829 | 60 | ||
XM_011538601.1 | XP_011536903.1 | 13 - 14 | 1866 | 60 | |
XM_011538602.1 | XP_011536904.1 | 14 - 15 | 1999 | 60 | |
XM_011538603.1 | XP_011536905.1 | 14 - 15 | 2017 | 60 | |
XM_011538604.1 | XP_011536906.1 | 14 - 15 | 1973 | 60 | |
XM_017019707.1 | XP_016875196.1 | 14 - 15 | 1999 | 60 | |
XM_017019708.1 | XP_016875197.1 | 12 - 13 | 1994 | 60 | |
XM_017019709.1 | XP_016875198.1 | 12 - 13 | 1994 | 60 | |
GenBank mRNA | AB040969.1 | - | 13 - 14 | 1829 | 60 |
AF370415.1 | - | 13 - 14 | 1837 | 60 | |
AK092788.1 | - | 12 - 13 | 1676 | 60 | |
AK122773.1 | - | 12 - 13 | 1593 | 60 | |
AK315578.1 | - | 13 - 14 | 1867 | 60 | |
AL136895.1 | - | 13 - 14 | 1844 | 60 | |
AY358397.1 | - | 13 - 14 | 1841 | 60 | |
AY563137.1 | - | 13 - 14 | 1839 | 60 | |
BC003177.1 | - | 13 - 14 | 1839 | 60 | |
BX538208.1 | - | 3 - 4 | 338 | 60 | |
CR533567.1 | - | 12 - 13 | 1788 | 60 |