The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020898.2 | NP_065949.1 | 8 - 9 | 1152 | 83 |
NR_026554.1 | 7 - 8 | 1049 | 83 | ||
XM_011538601.1 | XP_011536903.1 | 8 - 9 | 1086 | 83 | |
XM_011538602.1 | XP_011536904.1 | 9 - 10 | 1219 | 83 | |
XM_011538603.1 | XP_011536905.1 | 9 - 10 | 1237 | 83 | |
XM_011538604.1 | XP_011536906.1 | 9 - 10 | 1193 | 83 | |
XM_017019707.1 | XP_016875196.1 | 9 - 10 | 1219 | 83 | |
XM_017019708.1 | XP_016875197.1 | 7 - 8 | 1214 | 83 | |
XM_017019709.1 | XP_016875198.1 | 7 - 8 | 1214 | 83 | |
GenBank mRNA | AB040969.1 | - | 8 - 9 | 1049 | 83 |
AF370415.1 | - | 8 - 9 | 1057 | 83 | |
AK027881.1 | - | 8 - 9 | 1086 | 83 | |
AK092788.1 | - | 7 - 8 | 896 | 83 | |
AK294424.1 | - | 8 - 9 | 1060 | 83 | |
AK315578.1 | - | 8 - 9 | 1087 | 83 | |
AL136895.1 | - | 8 - 9 | 1064 | 83 | |
AY211909.1 | - | 4 - 5 | 571 | 83 | |
AY358397.1 | - | 8 - 9 | 1061 | 83 | |
AY563137.1 | - | 8 - 9 | 1059 | 83 | |
BC003177.1 | - | 8 - 9 | 1059 | 83 | |
CR533567.1 | - | 7 - 8 | 1008 | 83 |