Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGRF1
Entrez Gene ID
5923
Gene Name
Ras protein specific guanine nucleotide releasing factor 1
Gene Aliases
CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
Chromosome Location
Chr.15: 78959947 - 79090873 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145648.1 NP_001139120.1 22 - 23 3585 133
NM_002891.4 NP_002882.3 23 - 24 3633 133
NM_153815.2 NP_722522.1 9 - 10 2079 133
XM_011521866.2 XP_011520168.1 23 - 24 3533 133
XM_011521867.2 XP_011520169.1 23 - 24 3533 133
XM_017022455.1 XP_016877944.1 23 - 24 3524 133
XM_017022456.1 XP_016877945.1 22 - 23 3494 133
XM_017022457.1 XP_016877946.1 22 - 23 3494 133
XM_017022458.1 XP_016877947.1 22 - 23 3485 133
XM_017022459.1 XP_016877948.1 22 - 23 3494 133
GenBank mRNA AK226101.1 - 21 - 22 2869 133
AK309047.1 - 21 - 22 3016 133
BC040275.1 - 22 - 23 3476 133
L26584.1 - 23 - 24 3264 133
M91815.1 - 2 - 3 126 133
S62035.1 - 9 - 10 2088 133

View Full Product Details