Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGRF1
Entrez Gene ID
5923
Gene Name
Ras protein specific guanine nucleotide releasing factor 1
Gene Aliases
CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
Chromosome Location
Chr.15: 78959947 - 79090873 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145648.1 NP_001139120.1 8 - 9 1628 72
NM_002891.4 NP_002882.3 8 - 9 1628 72
XM_011521866.2 XP_011520168.1 8 - 9 1528 72
XM_011521867.2 XP_011520169.1 8 - 9 1528 72
XM_017022455.1 XP_016877944.1 8 - 9 1528 72
XM_017022456.1 XP_016877945.1 8 - 9 1528 72
XM_017022457.1 XP_016877946.1 8 - 9 1528 72
XM_017022458.1 XP_016877947.1 8 - 9 1528 72
XM_017022459.1 XP_016877948.1 8 - 9 1528 72
GenBank mRNA AK226101.1 - 7 - 8 912 72
AK290135.1 - 8 - 9 1494 72
AK309047.1 - 7 - 8 1050 72
BC040275.1 - 8 - 9 1519 72
L26584.1 - 8 - 9 1253 72

View Full Product Details