Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 64470793 - 64693058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293274.1 NP_001280203.1 9 - 10 1767 89
NM_020925.2 NP_065976.2 10 - 11 1488 89
XM_011541862.1 XP_011540164.1 9 - 10 1763 89
XM_011541863.1 XP_011540165.1 10 - 11 1912 89
GenBank mRNA AB046793.1 - 8 - 9 1270 89
BC039301.1 - 10 - 11 1474 89
BX537603.1 - 9 - 10 1766 89

View Full Product Details