Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 64470793 - 64693058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293274.1 NP_001280203.1 18 - 19 2960 71
NM_020925.2 NP_065976.2 19 - 20 2681 71
XM_011541862.1 XP_011540164.1 18 - 19 2956 71
XM_011541863.1 XP_011540165.1 19 - 20 3105 71
GenBank mRNA AB046793.1 - 17 - 18 2463 71
AK022571.1 - 6 - 7 779 71
AK024256.1 - 9 - 10 1106 71
AK299361.1 - 5 - 6 524 71
AL833000.1 - 9 - 10 1130 71
BC039301.1 - 19 - 20 2667 71
BX537603.1 - 18 - 19 2959 71

View Full Product Details