The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195736.1 | NP_001182665.1 | 6 - 7 | 765 | 62 |
NM_001195737.1 | NP_001182666.1 | 6 - 7 | 699 | 62 | |
NM_001195738.1 | NP_001182667.1 | 6 - 7 | 679 | 62 | |
NM_001195740.1 | NP_001182669.1 | 5 - 6 | 603 | 62 | |
NM_001195741.1 | NP_001182670.1 | 5 - 6 | 600 | 62 | |
NM_152371.3 | NP_689584.2 | 6 - 7 | 711 | 62 | |
NR_036637.1 | 6 - 7 | 644 | 62 | ||
NR_036638.1 | 6 - 7 | 612 | 62 | ||
XM_006710354.3 | XP_006710417.1 | 6 - 7 | 699 | 62 | |
XM_011540664.1 | XP_011538966.1 | 6 - 7 | 755 | 62 | |
XM_011540665.1 | XP_011538967.1 | 6 - 7 | 723 | 62 | |
XM_011540666.1 | XP_011538968.1 | 6 - 7 | 703 | 62 | |
GenBank mRNA | AF425266.1 | - | 6 - 7 | 655 | 62 |
AK057027.1 | - | 6 - 7 | 696 | 62 | |
AK075273.1 | - | 6 - 7 | 631 | 62 | |
AK094901.1 | - | 5 - 6 | 513 | 62 | |
AK291908.1 | - | 6 - 7 | 637 | 62 | |
AK298926.1 | - | 5 - 6 | 554 | 62 | |
AK303504.1 | - | 6 - 7 | 671 | 62 | |
AK308034.1 | - | 6 - 7 | 621 | 62 | |
AK316243.1 | - | 5 - 6 | 513 | 62 | |
BC022547.1 | - | 6 - 7 | 651 | 62 | |
BI908668.1 | - | 6 - 7 | 531 | 62 | |
BQ709693.1 | - | - | 547 | 62 | |
CR749706.1 | - | 1 - 2 | 443 | 62 |