Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGKA
Entrez Gene ID
1606
Gene Name
diacylglycerol kinase alpha
Gene Aliases
DAGK, DAGK1, DGK-alpha
Chromosome Location
Chr.12: 55931076 - 55954023 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345.4 NP_001336.2 3 - 4 407 65
NM_201444.2 NP_958852.1 3 - 4 452 65
NM_201445.1 NP_958853.1 4 - 5 420 65
NM_201554.1 NP_963848.1 3 - 4 320 65
XM_005268688.2 XP_005268745.1 3 - 4 349 65
XM_005268689.2 XP_005268746.1 3 - 4 273 65
XM_005268690.2 XP_005268747.1 1 - 2 79 65
XM_011537991.2 XP_011536293.1 4 - 5 517 65
XM_011537993.2 XP_011536295.1 3 - 4 268 65
XM_011537995.2 XP_011536297.1 3 - 4 268 65
XM_017018900.1 XP_016874389.1 3 - 4 349 65
XM_017018901.1 XP_016874390.1 3 - 4 493 65
XM_017018902.1 XP_016874391.1 3 - 4 315 65
XM_017018903.1 XP_016874392.1 4 - 5 517 65
XM_017018904.1 XP_016874393.1 4 - 5 385 65
XM_017018905.1 XP_016874394.1 5 - 6 794 65
XM_017018906.1 XP_016874395.1 3 - 4 426 65
XM_017018907.1 XP_016874396.1 3 - 4 249 65
XM_017018908.1 XP_016874397.1 3 - 4 349 65
XM_017018909.1 XP_016874398.1 2 - 3 224 65
GenBank mRNA AF064769.1 - 3 - 4 329 65
AF064770.1 - 3 - 4 382 65
AF064771.1 - 3 - 4 354 65
AK122973.1 - 1 - 2 1760 65
AK226134.1 - 1 - 2 961 65
AK303320.1 - 3 - 4 345 65
AK309939.1 - 3 - 4 296 65
AK310600.1 - 3 - 4 270 65
AY930112.1 - 3 - 4 347 65
BC023523.2 - 4 - 5 343 65
BC031870.2 - 3 - 4 347 65
BM836118.1 - 3 - 4 407 65
BM910598.1 - - 401 65
BQ943057.1 - 3 - 4 382 65
X62535.1 - 2 - 3 246 65

View Full Product Details