Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DGKA
- Entrez Gene ID
-
1606
- Gene Name
- diacylglycerol kinase alpha
- Gene Aliases
-
DAGK, DAGK1, DGK-alpha
- Chromosome Location
-
Chr.12: 55931076 - 55954023 on Build GRCh38
View Full Product Details