The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142733.2 | NP_001136205.2 | 5 - 6 | 585 | 83 |
XM_017005736.1 | XP_016861225.1 | 5 - 6 | 935 | 83 | |
XM_017005737.1 | XP_016861226.1 | 5 - 6 | 899 | 83 | |
GenBank mRNA | AL832472.1 | - | 5 - 6 | 586 | 83 |