The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003802.2 | NP_003793.2 | 25 - 26 | 3341 | 134 |
GenBank mRNA | AF111782.2 | - | 25 - 26 | 3341 | 134 |