Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM98A
Entrez Gene ID
25940
Gene Name
family with sequence similarity 98 member A
Gene Aliases
-
Chromosome Location
Chr.2: 33583661 - 33599362 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015475.4 NP_056290.3 4 - 5 87
GenBank mRNA AK074867.1 - 4 - 5 603 90
AK096187.1 - 1 - 2 87
BC038581.1 - 1 - 2 87
BC060860.1 - 4 - 5 87
BC136768.1 - 4 - 5 87
BC136770.1 - 4 - 5 87
CR936766.1 - 4 - 5 87

View Full Product Details