Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM98A
Entrez Gene ID
25940
Gene Name
family with sequence similarity 98 member A
Gene Aliases
-
Chromosome Location
Chr.2: 33583661 - 33599362 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304538.1 NP_001291467.1 4 - 5 555 69
NM_015475.4 NP_056290.3 5 - 6 740 69
GenBank mRNA AK074867.1 - 5 - 6 676 69
AK300023.1 - 4 - 5 484 69
AK302233.1 - 1 - 2 163 69
BC038581.1 - 2 - 3 273 69
BC060860.1 - 5 - 6 653 69
BC136768.1 - 5 - 6 644 69
BC136770.1 - 5 - 6 644 69
CR936766.1 - 5 - 6 678 69

View Full Product Details