Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELL
Entrez Gene ID
8178
Gene Name
elongation factor for RNA polymerase II
Gene Aliases
C19orf17, ELL1, MEN, PPP1R68
Chromosome Location
Chr.19: 18442663 - 18522129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006532.3 NP_006523.1 1 - 2 211 76
XM_011528330.2 XP_011526632.2 1 - 2 213 76
GenBank mRNA AB208794.1 - 1 - 2 132 76
AK225481.1 - 1 - 2 150 76
BC049195.1 - 1 - 2 193 76
BI458899.1 - 1 - 2 215 76
U16282.1 - 1 - 2 151 76

View Full Product Details