Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELL
Entrez Gene ID
8178
Gene Name
elongation factor for RNA polymerase II
Gene Aliases
C19orf17, ELL1, MEN, PPP1R68
Chromosome Location
Chr.19: 18442663 - 18522129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006532.3 NP_006523.1 6 - 7 942 109
XM_017027335.1 XP_016882824.1 7 - 8 926 109
XM_017027336.1 XP_016882825.1 2 - 3 678 109
GenBank mRNA AB208794.1 - 7 - 8 923 109
AK225481.1 - 6 - 7 881 109
AK293105.1 - 6 - 7 875 109
BC049195.1 - 6 - 7 924 109
U16282.1 - 6 - 7 882 109

View Full Product Details