Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMDN2
Entrez Gene ID
151393
Gene Name
regulator of microtubule dynamics 2
Gene Aliases
BLOCK18, FAM82A, FAM82A1, PRO34163, PYST9371, RMD-2, RMD2, RMD4
Chromosome Location
Chr.2: 37920789 - 38068032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170791.2 NP_001164262.1 5 - 6 919 100
NM_001170792.2 NP_001164263.1 5 - 6 990 100
NM_001170793.2 NP_001164264.1 4 - 5 496 100
NM_001322211.1 NP_001309140.1 5 - 6 988 100
NM_001322212.1 NP_001309141.1 5 - 6 919 100
NM_144713.4 NP_653314.3 5 - 6 1465 100
XM_011532613.2 XP_011530915.1 5 - 6 1446 100
XM_011532614.2 XP_011530916.1 7 - 8 1577 100
XM_011532615.2 XP_011530917.2 7 - 8 1577 100
XM_011532616.2 XP_011530918.1 5 - 6 891 100
XM_011532617.2 XP_011530919.1 5 - 6 977 100
XM_011532618.2 XP_011530920.1 6 - 7 1101 100
XM_011532619.2 XP_011530921.1 5 - 6 984 100
XM_017003474.1 XP_016858963.1 7 - 8 1577 100
XM_017003475.1 XP_016858964.1 7 - 8 1577 100
XM_017003476.1 XP_016858965.1 5 - 6 891 100
XM_017003477.1 XP_016858966.1 5 - 6 983 100
XM_017003478.1 XP_016858967.1 5 - 6 891 100
XM_017003479.1 XP_016858968.1 7 - 8 1070 100
GenBank mRNA AF435956.1 - 4 - 5 476 100
AK057516.1 - 5 - 6 1447 100
AK095462.1 - 6 - 7 976 100
AY358269.1 - 5 - 6 919 100
BC024243.2 - 5 - 6 992 100

View Full Product Details