The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166460.1 | NP_001159932.1 | 2 - 3 | 344 | 111 |
GenBank mRNA | AK304538.1 | - | 2 - 3 | 302 | 111 |