Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Chromosome Location
Chr.2: 215360440 - 215436167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306129.1 NP_001293058.1 38 - 39 6421 98
NM_001306130.1 NP_001293059.1 37 - 38 6151 98
NM_001306131.1 NP_001293060.1 36 - 37 5878 98
NM_001306132.1 NP_001293061.1 36 - 37 5878 98
NM_002026.3 NP_002017.1 37 - 38 6146 98
NM_212474.2 NP_997639.1 36 - 37 5876 98
NM_212476.2 NP_997641.1 36 - 37 5876 98
NM_212478.2 NP_997643.1 37 - 38 6146 98
NM_212482.2 NP_997647.1 38 - 39 6419 98
XM_005246397.1 XP_005246454.1 38 - 39 6518 98
XM_005246398.1 XP_005246455.1 38 - 39 6518 98
XM_005246399.1 XP_005246456.1 38 - 39 6518 98
XM_005246401.1 XP_005246458.1 38 - 39 6518 98
XM_005246402.1 XP_005246459.1 38 - 39 6518 98
XM_005246403.1 XP_005246460.1 37 - 38 6248 98
XM_005246404.1 XP_005246461.1 37 - 38 6245 98
XM_005246405.1 XP_005246462.1 37 - 38 6248 98
XM_005246406.1 XP_005246463.1 37 - 38 6245 98
XM_005246407.1 XP_005246464.1 38 - 39 6518 98
XM_005246408.1 XP_005246465.1 37 - 38 6248 98
XM_005246409.1 XP_005246466.1 37 - 38 6245 98
XM_005246410.1 XP_005246467.1 37 - 38 6248 98
XM_005246411.1 XP_005246468.1 37 - 38 6248 98
XM_005246412.1 XP_005246469.1 36 - 37 5975 98
XM_005246414.1 XP_005246471.1 37 - 38 6245 98
XM_005246416.1 XP_005246473.1 36 - 37 5975 98
XM_017003692.1 XP_016859181.1 38 - 39 6518 98
XM_017003693.1 XP_016859182.1 37 - 38 6248 98
XM_017003694.1 XP_016859183.1 37 - 38 6248 98
XM_017003695.1 XP_016859184.1 36 - 37 5975 98
GenBank mRNA AB191261.1 - 36 - 37 5876 98
AB209287.1 - 13 - 14 1849 98
AB209840.1 - 36 - 37 5868 98
AF130095.1 - 2 - 3 442 98
AJ320525.1 - 4 - 5 519 98
AJ320526.1 - 4 - 5 519 98
AJ320527.1 - 4 - 5 519 98
AK297733.1 - 3 - 4 368 98
AK300246.1 - 13 - 14 2031 98
AK300458.1 - 16 - 17 2497 98
AL832202.1 - 36 - 37 5877 98
AL832771.1 - 36 - 37 5877 98
BC117176.1 - 36 - 37 5856 98
BC143754.1 - 36 - 37 5856 98
BC143763.1 - 36 - 37 5856 98
BX537590.1 - 38 - 39 6421 98
BX538017.1 - 37 - 38 6147 98
BX538018.1 - 37 - 38 6147 98
BX640608.1 - 36 - 37 5876 98
BX640638.1 - 13 - 14 1919 98
BX640731.1 - 37 - 38 6147 98
BX640802.1 - 37 - 38 6145 98
BX640803.1 - 38 - 39 6420 98
BX640875.1 - 38 - 39 6420 98
BX640920.1 - 17 - 18 2582 98
BX640999.1 - 33 - 34 5395 98
BX641150.1 - 11 - 12 7248 98
BX649182.1 - 37 - 38 6145 98
CR749281.1 - 36 - 37 5915 98
CR749316.1 - 37 - 38 6148 98
CR749317.1 - 36 - 37 98
CR749666.1 - 17 - 18 2761 98
CR936623.1 - 14 - 15 2286 98
EF550130.1 - 8 - 9 1245 98
EF550131.1 - 8 - 9 1245 98
EF550132.1 - 7 - 8 975 98
EF550133.1 - 8 - 9 1245 98
EF550134.1 - 8 - 9 1245 98
EF550135.1 - 3 - 4 432 98
M10905.1 - 7 - 8 1103 98
M27589.1 - 1 - 2 40 98
U41724.1 - 1 - 2 126 98
U41850.1 - 1 - 2 126 98
U42455.1 - 1 - 2 126 98
U42593.1 - 9 - 10 1205 98
U42594.1 - 13 - 14 1915 98
U60067.1 - 6 - 7 760 98
U60068.1 - 6 - 7 760 98
X02761.1 - 37 - 38 5800 98

View Full Product Details