The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270890.1 | NP_001257819.1 | 6 - 7 | 944 | 105 |
NM_017645.4 | NP_060115.3 | 6 - 7 | 944 | 105 | |
XM_017014830.1 | XP_016870319.1 | 2 - 3 | 95 | 105 | |
GenBank mRNA | AF533709.1 | - | 6 - 7 | 1126 | 105 |
AF537091.1 | - | 6 - 7 | 906 | 105 | |
AK022964.1 | - | 3 - 4 | 260 | 105 | |
AK056458.1 | - | 6 - 7 | 906 | 105 | |
AL832495.1 | - | 6 - 7 | 904 | 105 | |
BC026178.1 | - | 6 - 7 | 907 | 105 | |
BC065935.1 | - | 4 - 5 | 444 | 105 | |
BC111041.1 | - | 2 - 3 | 220 | 105 | |
BC113981.1 | - | 6 - 7 | 741 | 105 | |
BC114492.1 | - | 6 - 7 | 741 | 105 |