The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017645.4 | NP_060115.3 | 9 - 10 | 1357 | 161 |
XM_017014830.1 | XP_016870319.1 | 5 - 6 | 508 | 161 | |
GenBank mRNA | AB046794.1 | - | 2 - 3 | 213 | 161 |
AF533709.1 | - | 9 - 10 | 1539 | 161 | |
AF537091.1 | - | 9 - 10 | 1319 | 161 | |
AK022964.1 | - | 6 - 7 | 673 | 161 | |
AK023068.1 | - | 2 - 3 | 276 | 161 | |
AK056458.1 | - | 9 - 10 | 1319 | 161 | |
AL832495.1 | - | 9 - 10 | 1317 | 161 | |
BC026178.1 | - | 9 - 10 | 1320 | 161 | |
BC065935.1 | - | 7 - 8 | 857 | 161 | |
BC071625.1 | - | 2 - 3 | 316 | 161 | |
BC111041.1 | - | 5 - 6 | 633 | 161 | |
BC113981.1 | - | 9 - 10 | 1154 | 161 | |
BC114492.1 | - | 9 - 10 | 1154 | 161 |