Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOD2
Entrez Gene ID
64127
Gene Name
nucleotide binding oligomerization domain containing 2
Gene Aliases
ACUG, BLAU, CARD15, CD, CLR16.3, IBD1, NLRC2, NOD2B, PSORAS1
Chromosome Location
Chr.16: 50693581 - 50733077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293557.1 NP_001280486.1 4 - 5 2608 88
NM_022162.2 NP_071445.1 5 - 6 2648 88
XM_005256084.3 XP_005256141.1 5 - 6 2552 88
XM_006721242.3 XP_006721305.1 5 - 6 2552 88
XM_011523259.2 XP_011521561.1 3 - 4 2364 88
XM_017023535.1 XP_016879024.1 3 - 4 3922 88
XM_017023536.1 XP_016879025.1 4 - 5 2079 88
XM_017023537.1 XP_016879026.1 3 - 4 1973 88
XM_017023538.1 XP_016879027.1 4 - 5 2074 88
GenBank mRNA AF178930.1 - 5 - 6 2648 88
AY187233.1 - 2 - 3 261 88
AY187234.1 - 2 - 3 289 88
AY187241.1 - 2 - 3 261 88

View Full Product Details