Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJC1
Entrez Gene ID
64215
Gene Name
DnaJ heat shock protein family (Hsp40) member C1
Gene Aliases
DNAJL1, ERdj1, HTJ1, MTJ1
Chromosome Location
Chr.10: 21756548 - 22003768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022365.3 NP_071760.2 8 - 9 1268 63
XM_011519614.2 XP_011517916.1 8 - 9 1315 63
GenBank mRNA AF427142.1 - 1 - 2 144 63
AK026062.1 - 5 - 6 541 63
AK027263.1 - 8 - 9 1088 63
AK308369.1 - 8 - 9 64
AY225122.1 - 8 - 9 981 63
BC030955.1 - 4 - 5 451 63
BC110894.1 - 8 - 9 1268 63

View Full Product Details