Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG2
Entrez Gene ID
1740
Gene Name
discs large MAGUK scaffold protein 2
Gene Aliases
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location
Chr.11: 83455009 - 85628534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142699.1 NP_001136171.1 16 - 17 1884 84
NM_001142700.1 NP_001136172.1 8 - 9 973 84
NM_001206769.1 NP_001193698.1 10 - 11 1568 84
NM_001300983.1 NP_001287912.1 11 - 12 1560 84
NM_001364.3 NP_001355.2 11 - 12 1560 84
XM_005273810.4 XP_005273867.1 10 - 11 1203 84
XM_005273811.4 XP_005273868.1 10 - 11 1203 84
XM_011544778.2 XP_011543080.1 17 - 18 3175 84
XM_011544780.2 XP_011543082.1 17 - 18 3175 84
XM_011544782.2 XP_011543084.1 12 - 13 1646 84
XM_011544788.2 XP_011543090.1 9 - 10 1047 84
XM_017017254.1 XP_016872743.1 17 - 18 3175 84
XM_017017255.1 XP_016872744.1 16 - 17 3136 84
XM_017017256.1 XP_016872745.1 16 - 17 2665 84
XM_017017257.1 XP_016872746.1 15 - 16 3597 84
XM_017017258.1 XP_016872747.1 14 - 15 2773 84
XM_017017259.1 XP_016872748.1 15 - 16 2590 84
XM_017017260.1 XP_016872749.1 14 - 15 3522 84
XM_017017261.1 XP_016872750.1 16 - 17 3019 84
XM_017017262.1 XP_016872751.1 10 - 11 1628 84
XM_017017263.1 XP_016872752.1 12 - 13 1646 84
XM_017017264.1 XP_016872753.1 11 - 12 1823 84
XM_017017265.1 XP_016872754.1 11 - 12 1820 84
XM_017017266.1 XP_016872755.1 10 - 11 1626 84
XM_017017267.1 XP_016872756.1 11 - 12 1728 84
XM_017017268.1 XP_016872757.1 11 - 12 1549 84
XM_017017269.1 XP_016872758.1 11 - 12 2250 84
XM_017017270.1 XP_016872759.1 12 - 13 1646 84
XM_017017271.1 XP_016872760.1 11 - 12 1823 84
XM_017017272.1 XP_016872761.1 11 - 12 1820 84
XM_017017273.1 XP_016872762.1 9 - 10 1479 84
XM_017017274.1 XP_016872763.1 11 - 12 1728 84
XM_017017275.1 XP_016872764.1 9 - 10 1471 84
XM_017017276.1 XP_016872765.1 11 - 12 2248 84
XM_017017277.1 XP_016872766.1 10 - 11 1472 84
XM_017017278.1 XP_016872767.1 9 - 10 1472 84
XM_017017279.1 XP_016872768.1 11 - 12 1490 84
XM_017017280.1 XP_016872769.1 11 - 12 2167 84
XM_017017281.1 XP_016872770.1 10 - 11 1667 84
XM_017017282.1 XP_016872771.1 10 - 11 1203 84
XM_017017283.1 XP_016872772.1 10 - 11 1472 84
XM_017017284.1 XP_016872773.1 9 - 10 1048 84
XM_017017285.1 XP_016872774.1 10 - 11 2094 84
XM_017017286.1 XP_016872775.1 7 - 8 912 84
GenBank mRNA AB209252.1 - 11 - 12 1535 84
AK126776.1 - 8 - 9 973 84
AK127150.1 - 10 - 11 1566 84
AK294375.1 - 10 - 11 1091 84
AK295094.1 - 11 - 12 1560 84
AK295624.1 - 10 - 11 1183 84
AK299797.1 - 11 - 12 1318 84
CR749820.1 - 16 - 17 1884 84
KJ535093.1 - 16 - 17 1767 84
U32376.1 - 11 - 12 1533 84

View Full Product Details