The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005366.1 | NP_001005366.1 | 5 - 6 | 893 | 97 |
NM_032590.4 | NP_115979.3 | 5 - 6 | 683 | 97 | |
XM_005253955.3 | XP_005254012.1 | 5 - 6 | 893 | 97 | |
XM_005253956.3 | XP_005254013.1 | 5 - 6 | 490 | 97 | |
XM_011538867.2 | XP_011537169.1 | 5 - 6 | 1936 | 97 | |
XM_011538868.2 | XP_011537170.1 | 5 - 6 | 1932 | 97 | |
XM_011538869.2 | XP_011537171.1 | 5 - 6 | 1934 | 97 | |
GenBank mRNA | AJ459424.1 | - | 5 - 6 | 655 | 97 |
AK097933.1 | - | 3 - 4 | 303 | 97 | |
AK127328.1 | - | 5 - 6 | 893 | 97 | |
AL133572.1 | - | 1 - 2 | 196 | 97 | |
BC115379.1 | - | 4 - 5 | 543 | 97 | |
BC115380.1 | - | 5 - 6 | 622 | 97 | |
BX648928.1 | - | 5 - 6 | 685 | 97 |